Learn More
Medchemexpress LLC Mivavotinib monohydrochloride | 1952251-28-3 | 99.5% | 380.85 | C17H22ClFN6O | 10 MG
SDP
Catalog No. 5000211719
Encompass_Preferred
Mivavotinib monohydrochloride is the hydrochloride salt of mivavotinib, a potent, selective, reversible dual inhibitor of spleen tyrosine kinase (SYK) and fms-related tyrosine kinase 3 (FLT3) used for preclinical research. Supplied as a high-purity research chemical, it is used to probe SYK/FLT3 signaling and antitumor activity.
  • Potent dual SYK and FLT3 inhibition, with reported IC50 values of 3.2 nM and 4.6 nM.
  • High purity suitable for research (data sheet reported 99.50%).
  • Available in small milligram pack sizes for preclinical experiments.
  • Monohydrochloride salt form for improved stability and solubility compared with the free base.
  • Molecular weight 380.85 and formula C17H22ClFN6O for accurate dosing calculations.
Catalog No. 50-002-11719 Supplier Medchemexpress LLC Supplier No. HY100867A10MG
Mivavotinib monohydrochloride is the hydrochloride salt of mivavotinib, a potent, selective, reversible dual inhibitor of spleen tyrosine kinase (SYK) and fms-related tyrosine kinase 3 (FLT3) used for preclinical research. Supplied as a high-purity research chemical, it is used to probe SYK/FLT3 signaling and antitumor activity.
  • Potent dual SYK and FLT3 inhibition, with reported IC50 values of 3.2 nM and 4.6 nM.
  • High purity suitable for research (data sheet reported 99.50%).
  • Available in small milligram pack sizes for preclinical experiments.
  • Monohydrochloride salt form for improved stability and solubility compared with the free base.
  • Molecular weight 380.85 and formula C17H22ClFN6O for accurate dosing calculations.