Learn More
Medchemexpress LLC Mivavotinib monohydrochloride | 1952251-28-3 | 99.9% | 380.85 g/mol | C17H22ClFN6O | 5 MG
SDP
Catalog No. 5000211283
Encompass_Preferred
Mivavotinib monohydrochloride is an investigational small-molecule dual inhibitor of spleen tyrosine kinase (SYK) and fms related tyrosine kinase 3 (FLT3), supplied as a white to off-white solid for research use. The monohydrochloride salt displays low-nanomolar inhibitory activity against SYK and FLT3 and is provided with high purity for preclinical studies.
  • Dual SYK/FLT3 inhibitor for preclinical research.
  • High purity (99.9%).
  • White to off-white solid formulation.
  • Provided as a monohydrochloride salt for improved stability.
  • Low-nanomolar potency against target kinases.
  • Store at 4°C and protect from moisture for long-term stability.
Catalog No. 50-002-11283 Supplier Medchemexpress LLC Supplier No. HY100867A5MG
Mivavotinib monohydrochloride is an investigational small-molecule dual inhibitor of spleen tyrosine kinase (SYK) and fms related tyrosine kinase 3 (FLT3), supplied as a white to off-white solid for research use. The monohydrochloride salt displays low-nanomolar inhibitory activity against SYK and FLT3 and is provided with high purity for preclinical studies.
  • Dual SYK/FLT3 inhibitor for preclinical research.
  • High purity (99.9%).
  • White to off-white solid formulation.
  • Provided as a monohydrochloride salt for improved stability.
  • Low-nanomolar potency against target kinases.
  • Store at 4°C and protect from moisture for long-term stability.