Learn More
Medchemexpress LLC HY-100242 10mg Medchemexpress, Mivotilate CAS:130112-42-4 Purity:>98%
SDP
Catalog No. 501967718
Encompass_Preferred
Medchemexpress, HY-100242 10mg Mivotilate CAS:130112-42-4 Mivotilate is a nontoxic, potent activator of the aryl hydrocarbon receptor (AhR), and acts as a hepatoprotective agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7718 Supplier Medchemexpress LLC Supplier No. HY10024210MG
Medchemexpress, HY-100242 10mg Mivotilate CAS:130112-42-4 Mivotilate is a nontoxic, potent activator of the aryl hydrocarbon receptor (AhR), and acts as a hepatoprotective agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.