Learn More
Medchemexpress LLC Mivotilate | 130112-42-4 | MFCD02752797 | 99.6% | 330.45 g/mol | C12H14N2O3S3 | 100 MG
SDP
Catalog No. 5000215528
Encompass_Preferred
Mivotilate is a small-molecule activator of the aryl hydrocarbon receptor (AhR) with reported hepatoprotective activity. Provided as a research reagent and analytical standard, it is supplied as a solid and as a DMSO solution for biochemical and cell-based assays.
  • Activates aryl hydrocarbon receptor (AhR).
  • Described as nontoxic in supplier literature.
  • High purity (99.6%).
  • Molecular weight 330.45 g/mol.
  • Available as solid and DMSO solution for flexibility in use.
  • Suitable for biochemical and in vitro studies.
Catalog No. 50-002-15528 Supplier Medchemexpress LLC Supplier No. HY100242100MG
Mivotilate is a small-molecule activator of the aryl hydrocarbon receptor (AhR) with reported hepatoprotective activity. Provided as a research reagent and analytical standard, it is supplied as a solid and as a DMSO solution for biochemical and cell-based assays.
  • Activates aryl hydrocarbon receptor (AhR).
  • Described as nontoxic in supplier literature.
  • High purity (99.6%).
  • Molecular weight 330.45 g/mol.
  • Available as solid and DMSO solution for flexibility in use.
  • Suitable for biochemical and in vitro studies.