Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mivotilate | 130112-42-4 | MFCD02752797 | 99.6% | 330.45 g/mol | C12H14N2O3S3 | 50 MG
SDP

Catalog No. 5000215778
Encompass_Preferred

Mivotilate (YH439) is a small-molecule activator of the aryl hydrocarbon receptor (AhR) with reported hepatoprotective activity. It induces cytochromes P4501A1/2 and has been shown in vivo to reduce CYP2E1-mediated NDMA demethylase activity and lower CYP2E1 protein levels.

  • Potent activator of the aryl hydrocarbon receptor (AhR).
  • Demonstrated hepatoprotective activity in vivo.
  • Induces CYP1A1/2 and downregulates CYP2E1 transcription.
  • High purity suitable for research applications.
  • Available in multiple package sizes for experimental flexibility.
  • Molecular weight 330.45 g/mol; molecular formula C12H14N2O3S3.

Catalog No. 50-002-15778 Supplier Medchemexpress LLC Supplier No. HY10024250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mivotilate (YH439) is a small-molecule activator of the aryl hydrocarbon receptor (AhR) with reported hepatoprotective activity. It induces cytochromes P4501A1/2 and has been shown in vivo to reduce CYP2E1-mediated NDMA demethylase activity and lower CYP2E1 protein levels.

  • Potent activator of the aryl hydrocarbon receptor (AhR).
  • Demonstrated hepatoprotective activity in vivo.
  • Induces CYP1A1/2 and downregulates CYP2E1 transcription.
  • High purity suitable for research applications.
  • Available in multiple package sizes for experimental flexibility.
  • Molecular weight 330.45 g/mol; molecular formula C12H14N2O3S3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.