Learn More
Medchemexpress LLC Mivotilate (standard) | 130112-42-4 | 99.6% | 330.45 g/mol | C12H14N2O3S3 | 25 MG
SDP
Catalog No. 5000215510
Encompass_Preferred
Mivotilate is a research-grade small-molecule activator of the aryl hydrocarbon receptor (AhR) with reported hepatoprotective activity. It is supplied as a purified analytical standard for laboratory use and is intended for in vitro research and analytical applications.
  • Potent, nontoxic activator of the aryl hydrocarbon receptor.
  • Reported hepatoprotective activity in preclinical studies.
  • High purity suitable for analytical and research use.
  • Molecular weight 330.45 g/mol; formula C12H14N2O3S3.
  • Available in small, research-oriented pack sizes.
Catalog No. 50-002-15510 Supplier Medchemexpress LLC Supplier No. HY10024225MG
Mivotilate is a research-grade small-molecule activator of the aryl hydrocarbon receptor (AhR) with reported hepatoprotective activity. It is supplied as a purified analytical standard for laboratory use and is intended for in vitro research and analytical applications.
  • Potent, nontoxic activator of the aryl hydrocarbon receptor.
  • Reported hepatoprotective activity in preclinical studies.
  • High purity suitable for analytical and research use.
  • Molecular weight 330.45 g/mol; formula C12H14N2O3S3.
  • Available in small, research-oriented pack sizes.