Learn More
Research Products International Corp Mix & Go! Competent Cells-JM109, 10 x 100 ul, 10 Tubes
SDP
Catalog No. 501977696
Encompass
The Mix & Go! E. coli strains are premade, chemically competent cells for simple and highly efficient DNA transformation. Mix & Go! E. coli cells are made chemically competent by a method that completely eliminates the need for heat shocking and related procedures. For transformation, simply mix DNA with cells and then spread onto solid medium − Mix & Go! The premade Mix & Go! competent cells are highly efficient (> 108 transformants/ µg pUC19) and can be used for cloning, sub-cloning, PCR fragment cloning, library construction, etc.
Catalog No. 50-197-7696 Supplier Research Products International Corp Supplier No. ZT3003
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Mix & Go! E. coli strains are premade, chemically competent cells for simple and highly efficient DNA transformation. Mix & Go! E. coli cells are made chemically competent by a method that completely eliminates the need for heat shocking and related procedures. For transformation, simply mix DNA with cells and then spread onto solid medium − Mix & Go! The premade Mix & Go! competent cells are highly efficient (> 108 transformants/ µg pUC19) and can be used for cloning, sub-cloning, PCR fragment cloning, library construction, etc.