Learn More
Research Products International Corp Mix & Go! Competent Cells-Zymo 5α, 10 x 100 ul, 10 tubes
SDP
Catalog No. 501977697
Encompass
The Mix & Go! Competent Cells - DH5 Alpha are premade E. coli DH5 alpha competent cells for simple and highly efficient DNA transformation. These DH5 alpha competent cells are made chemically competent by a method that completely eliminates the need for heat shocking and related procedures. For transformation, simply mix DNA with cells and then spread onto solid medium − Mix & Go! The premade Mix & Go! competent cells are highly efficient (> 108 transformants/µg pUC19) and can be used for cloning, sub-cloning, PCR fragment cloning, library construction, etc.
Simple 20 Second Transformation: No heat shock! Just add DNA and spread on plate.
High Transformation Efficiencies: Achieve 108 - 109 per µg of plasmid DNA.
Versatile: Excellent for general cloning, blue-white screening, and plasmid isolation.
Catalog No. 50-197-7697 Supplier Research Products International Corp Supplier No. ZT3007
The Mix & Go! Competent Cells - DH5 Alpha are premade E. coli DH5 alpha competent cells for simple and highly efficient DNA transformation. These DH5 alpha competent cells are made chemically competent by a method that completely eliminates the need for heat shocking and related procedures. For transformation, simply mix DNA with cells and then spread onto solid medium − Mix & Go! The premade Mix & Go! competent cells are highly efficient (> 108 transformants/µg pUC19) and can be used for cloning, sub-cloning, PCR fragment cloning, library construction, etc.
Simple 20 Second Transformation: No heat shock! Just add DNA and spread on plate.
High Transformation Efficiencies: Achieve 108 - 109 per µg of plasmid DNA.
Versatile: Excellent for general cloning, blue-white screening, and plasmid isolation.