Learn More
Zymo Research Corporation Mix & Go! XJb Autolysis Competent Cells, 1 ml 500x L-Arabinose, 10 x 100 µl
SDP
Catalog No. 502037284
Encompass_Preferred
Additionally, it does not require use of any potentially interferingcomponents such as detergents, commonly found in various lytic buffers.Highlights• Simple and controlled autolysis of E. coli• Strains can by lysed in minutes after harvesting• Method compatible with most buffer systems• Ideal for protein expression and purification, also applicable for extraction of nucleic acids• Scale up to lyse more samples without increase in time or labor
Catalog No. 50-203-7284 Supplier Zymo Research Corporation Supplier No. T3041
Additionally, it does not require use of any potentially interferingcomponents such as detergents, commonly found in various lytic buffers.Highlights• Simple and controlled autolysis of E. coli• Strains can by lysed in minutes after harvesting• Method compatible with most buffer systems• Ideal for protein expression and purification, also applicable for extraction of nucleic acids• Scale up to lyse more samples without increase in time or labor