Learn More
Biotium Mix-n-Stain™ FITC Antibody Labeling Kit, 1x(20-50ug) labeling
Catalog No. NC1675931
Encompass_Preferred
Mix-n-Stain™ FITC antibody labeling kits are a revolutionary antibody labeling technology that allows you to label your antibody with fluorescein (FITC) in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody.
Catalog No. NC1675931 Supplier Biotium Supplier No. 92295
Mix-n-Stain™ FITC antibody labeling kits are a revolutionary antibody labeling technology that allows you to label your antibody with fluorescein (FITC) in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody.