Learn More
Sigma Aldrich Fine Chemicals Biosciences Mix-n-StainTM CfTM 405S An
Catalog No. 502901612
Encompass
This labeling kit is for labeling 50-100 mug antibody with the blue fluorescent dye Cf405S. Mix-n-Stain Cf dye antibody labeling kits are a revolutionary antibody labeling technology that permits one to label antibodies with our superior Cf dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Cf405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result flow cytometry analysis using Cf405S results in much brighter signal than that using Alexa fluor(R) 405.
Catalog No. 50-290-1612 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MX405SS1001KT
Add to Cart
Add to Cart
This labeling kit is for labeling 50-100 mug antibody with the blue fluorescent dye Cf405S. Mix-n-Stain Cf dye antibody labeling kits are a revolutionary antibody labeling technology that permits one to label antibodies with our superior Cf dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Cf405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result flow cytometry analysis using Cf405S results in much brighter signal than that using Alexa fluor(R) 405.