Learn More
Sorbent Technologies Inc Special Mixed Layer of Silica with Strongly Acidic Cation Exchange Resin, poly backed, 200um, 20x20cm, 25pk
SDP
Catalog No. 501138522
Encompass_Preferred
Special mixed layer of silica and a strongly acidic cation exchanger coated to polyester plates. The coating thickness is 200um and dimensions are 20x20cm. There are 25 plates per pack. Applications include: Amino acids, e.g., in protein and peptide hydrolysates, in seeds and fodder, in biological fluids; for racemate separation in peptide syntheses, for the separation of nucleic acid hydrolyzates, amino sugars, amino acids, antibiotics, inorganic phosphates, cations and other compounds with ionic groups.
Catalog No. 50-113-8522 Supplier Sorbent Technologies Inc Supplier No. 1224026
Special mixed layer of silica and a strongly acidic cation exchanger coated to polyester plates. The coating thickness is 200um and dimensions are 20x20cm. There are 25 plates per pack. Applications include: Amino acids, e.g., in protein and peptide hydrolysates, in seeds and fodder, in biological fluids; for racemate separation in peptide syntheses, for the separation of nucleic acid hydrolyzates, amino sugars, amino acids, antibiotics, inorganic phosphates, cations and other compounds with ionic groups.