Learn More
Julabo USA Inc Each, test tube rack tray for SW22/SW23 shaking water baths
SDP
Catalog No. 507534082
Encompass_Preferred
Bath accessories, Julabo, Basic tray for assembling max. 4 test tube racks. Basic tray for assembling max. 4 racks 8970380, 8970381, 8970382, 8970383 8970344, 8970345, 8970346 or 8970347. For use with shaking water baths SW22 or SW23.
Catalog No. 50-753-4082 Supplier Julabo USA Inc Supplier No. 8970369
Bath accessories, Julabo, Basic tray for assembling max. 4 test tube racks. Basic tray for assembling max. 4 racks 8970380, 8970381, 8970382, 8970383 8970344, 8970345, 8970346 or 8970347. For use with shaking water baths SW22 or SW23.