Learn More
2Mag USA 2mag MIXcontrol eco DINrail 0-10 V Control Unit
SDP
Catalog No. NC2736778
Encompass
Price optimized universal control unit with DINrail mounted interface (i.e., electrical cabinet) designed for use with/connection of one (1) 2mag MIXdrive Stirring Drive. Ideal for small to medium stirring volumes. Features stirring speed range from 120 to 1,200 rpm and stirring power up to 10 watts.
Catalog No. NC2736778 Supplier 2Mag USA Supplier No. MF 90150
Price optimized universal control unit with DINrail mounted interface (i.e., electrical cabinet) designed for use with/connection of one (1) 2mag MIXdrive Stirring Drive. Ideal for small to medium stirring volumes. Features stirring speed range from 120 to 1,200 rpm and stirring power up to 10 watts.