Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mixed Calibration Standard 4, ICP, Ricca Chemical
SDP

Catalog No. p-3339943 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RMXCL4-100 100 mL Natural LDPE Bottle
RMXCL4-500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RMXCL4-100 Supplier Ricca Chemical Company Supplier No. RMXCL4100
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Mixed Calibration 4
Packaging Natural LDPE Bottle
CAS 7697-37-2, 7732-18-5
Odor Odorless
Color Colorless
Melting Point Approximately 0°C
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.