Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mixed Calibration Standard 3A, ICP, Rev 3.3, Ricca Chemical
SDP

Catalog No. RMXCL3A500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RMXCL3A-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RMXCL3A-500 Supplier Ricca Chemical Company Supplier No. RMXCL3A500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Mixed Calibration Standard 3A
Packaging Natural Poly Bottle
Name Note Rev 3.3
CAS 7732-18-5, 7722-76-1, 7440-48-4, 7440-62-2, 7697-37-2, 7732-18-5
CAS Min % 91.0, 6.81, 0.03, 0.0016, 0.0015
CAS Max % 95.0, 7.09, 0.03, 0.0016, 0.0015
Odor Odorless
Color Colorless
pH Acidic
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 500 mL
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.