Learn More
Brainxell Inc Mixed Cortical Neurons Kit, 5M Viable Cells/Kit- Standard (BX-0500)
SDP
Catalog No. NC3001383
Encompass
Origin: Human iPSC lineProducts5M Viable Cells/Kit- Standard (BX-0500)Contents: 1 vial Cortical Glutamatergic Neurons (>4M), 1 vial Cortical GABAergic Neurons (>1M), and corresponding BrainXell SupplementsCryopreserved
Catalog No. NC3001383 Supplier Brainxell Inc Supplier No. BX0500305M
Origin: Human iPSC lineProducts5M Viable Cells/Kit- Standard (BX-0500)Contents: 1 vial Cortical Glutamatergic Neurons (>4M), 1 vial Cortical GABAergic Neurons (>1M), and corresponding BrainXell SupplementsCryopreserved