Learn More
Mixed Indicator Solution, Methyl Red-Methylene Blue, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Mixed Indicator Solution |
| Name Note | Methyl Red-Methylene Blue |
| CAS | 64-17-5, 67-56-1, 845-10-3, 7220-79-3 |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Nonaqueous Solution |
| Color | Dark Green |
| Quantity | 1 L |
| Physical Form | Liquid |
| Grade | Indicator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.