Learn More
Hach Company Mixed-Parameter Standard, NIST, Wastewater, Low Range
Catalog No. 501997739 Shop All Hach Company Products
Encompass
Mixed-Parameter quality control standard for wastewater parameters, low range. Components of the standard solution (with the exception of ammonia) are traceable to SRM from NIST.COD – 50 mg/LPO4-P – 1.0 mg/LNH3-N – 1.0 mg/LNO3-N – 6.0 mg/LTN – 7.0 mg/LApplicable for TNTplus vial tests- TNT821 COD, 3-150 mg/L O2- TNT843 Phosphate, 0.05-1.5 mg/L PO4-P- TNT830 Ammonia, 0.02-2.5 mg/L NH3-N- TNT835 Nitrate, 0.2-13.5 mg/L NO3-N- TNT826 Total Nitrogen, 1-16 mg/L TN
Catalog No. 50-199-7739 Supplier Hach Company Supplier No. LCA721
Mixed-Parameter quality control standard for wastewater parameters, low range. Components of the standard solution (with the exception of ammonia) are traceable to SRM from NIST.COD – 50 mg/LPO4-P – 1.0 mg/LNH3-N – 1.0 mg/LNO3-N – 6.0 mg/LTN – 7.0 mg/LApplicable for TNTplus vial tests- TNT821 COD, 3-150 mg/L O2- TNT843 Phosphate, 0.05-1.5 mg/L PO4-P- TNT830 Ammonia, 0.02-2.5 mg/L NH3-N- TNT835 Nitrate, 0.2-13.5 mg/L NO3-N- TNT826 Total Nitrogen, 1-16 mg/L TN