Learn More
Aqua Solutions MIXED SALT SOLUTION, DIL 500ML
SDP
Supplier:  Aqua Solutions 5916500ML
Encompass_Preferred
Mixed Salt Solution, Dilute in Mixed Alcohol Solvent ASTM D 3230
Catalog No. NC0951423
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov