Learn More
Waters Corp Waters Corporation Mixer, 250 μL, Titanium, Bio-Inert
Catalog No. 502039236 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 250 μL mixer is an optional component designed for the H-Class Bio Quaternary Solvent Manager. Made of titanium, it replaces the standard 100 μL mixer in applications requiring the smoothest baseline when using an optical detector.
  • Designed for H-Class Bio Quaternary Solvent Manager systems.
  • Features a 250 μL volume.
  • Constructed from titanium.
  • Replaces standard 100 μL mixers for enhanced performance.
  • Recommended for monitoring low UV wavelengths (below approximately 225 nm) in the presence of UV-absorbing mobile phases.
  • Provides a smoother baseline for optical detection.
  • Can be used in mass spectrometry systems when methods are intended for both MS and UV detection.
Catalog No. 50-203-9236 Supplier Waters Corp Supplier No. 205000737
This 250 μL mixer is an optional component designed for the H-Class Bio Quaternary Solvent Manager. Made of titanium, it replaces the standard 100 μL mixer in applications requiring the smoothest baseline when using an optical detector.
  • Designed for H-Class Bio Quaternary Solvent Manager systems.
  • Features a 250 μL volume.
  • Constructed from titanium.
  • Replaces standard 100 μL mixers for enhanced performance.
  • Recommended for monitoring low UV wavelengths (below approximately 225 nm) in the presence of UV-absorbing mobile phases.
  • Provides a smoother baseline for optical detection.
  • Can be used in mass spectrometry systems when methods are intended for both MS and UV detection.