Learn More
Waters Corp Mixer Assembly, 100 μL, Biocompatible, Zirconia Bead
Catalog No. 502390705 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This mixer assembly is a 100 μL biocompatible, packed bead mixer with zirconia beads. It serves as a replacement part and is specifically designed for the ACQUITY H-Class Bio Quaternary Solvent Manager, ensuring all wetted materials are bio-inert.
  • 100 μL volume
  • Biocompatible mixer assembly
  • Designed for the ACQUITY H-Class Bio Quaternary Solvent Manager
  • Standard packed bead mixer type
  • Features zirconia beads
  • All wetted materials are bio-inert
Catalog No. 50-239-0705 Supplier Waters Corp Supplier No. 700005258
This mixer assembly is a 100 μL biocompatible, packed bead mixer with zirconia beads. It serves as a replacement part and is specifically designed for the ACQUITY H-Class Bio Quaternary Solvent Manager, ensuring all wetted materials are bio-inert.
  • 100 μL volume
  • Biocompatible mixer assembly
  • Designed for the ACQUITY H-Class Bio Quaternary Solvent Manager
  • Standard packed bead mixer type
  • Features zirconia beads
  • All wetted materials are bio-inert