Learn More
Waters Corp Waters Mixer Assembly, 100μL, Packed Bead, Zirconia Beads, UPLC H-Class System
Catalog No. 50466627 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Designed for the UPLC H-Class System
  • Standard mixer type
  • 100μL volume
  • Contains packed zirconia beads
  • Intended for the Quaternary Solvent Manager (QSM)
  • Replacement part for maintenance as required
  • One unit per package
Catalog No. 50-466-627 Supplier Waters Corp Supplier No. 700005119
  • Designed for the UPLC H-Class System
  • Standard mixer type
  • 100μL volume
  • Contains packed zirconia beads
  • Intended for the Quaternary Solvent Manager (QSM)
  • Replacement part for maintenance as required
  • One unit per package