Learn More
HOLLAND APPLIED TECHNOLOGIES MIXER ASSEMBLY 1L
Catalog No. NC3640687
Encompass
  • Multi Inlet Vortex Mixer (MIVM) technology
  • Designed for rapid micromixing
  • Supports Flash NanoPrecipitation (FNP) applications
  • Able to manage up to four distinct inlet streams
  • Facilitates scalable production of uniform nanoparticles
  • Developed in collaboration with Princeton University
  • Efficient mixing for precise particle characteristics
  • 1L nominal volume/scale
  • Ideal for scientific and biotechnology applications
Catalog No. NC3640687 Supplier HOLLAND APPLIED TECHNOLOGIES Supplier No. MIVM1L KIT
  • Multi Inlet Vortex Mixer (MIVM) technology
  • Designed for rapid micromixing
  • Supports Flash NanoPrecipitation (FNP) applications
  • Able to manage up to four distinct inlet streams
  • Facilitates scalable production of uniform nanoparticles
  • Developed in collaboration with Princeton University
  • Efficient mixing for precise particle characteristics
  • 1L nominal volume/scale
  • Ideal for scientific and biotechnology applications