Learn More
Waters Corp Mixer Assembly, 250μL, Zirconia
Supplier:  Waters Corp 205000719
Encompass_Preferred
This replacement mixer assembly features a 250μL capacity and Zirconia construction. It is an optional mixer for the H-Class and H-Class PLUS Quaternary Solvent Managers.
  • Designed for UPLC systems.
  • 250μL capacity.
  • Constructed from Zirconia.
  • Optional component for H-Class and H-Class PLUS Quaternary Solvent Managers.
Catalog No. 50-239-2580