Learn More
Waters Corp Mixer Assembly, 250 μL, UPC2
Catalog No. 502390767 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a replacement mixer assembly with a capacity of 250 μL, designed for use in the UPC2 Binary Solvent Manager.
  • Replacement 250μL mixer assembly.
  • Used in UPC2 binary solvent manager.
Catalog No. 50-239-0767 Supplier Waters Corp Supplier No. 700008909
This product is a replacement mixer assembly with a capacity of 250 μL, designed for use in the UPC2 Binary Solvent Manager.
  • Replacement 250μL mixer assembly.
  • Used in UPC2 binary solvent manager.