Learn More
Cole-Parmer Koflo 1/2-40-3-12-2 Pipe Mixer, 316 stainless steel, 1/2" NPT(M), 12 elements
Catalog No. 50200207 Shop All Cole-Parmer Products
Encompass_Preferred
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
Catalog No. 50-200-207 Supplier Cole-Parmer Supplier No. 0466918
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.