Learn More
USA Scientific Inc Mixer HC ISO Rack 0.5 mL tube
Catalog No. 502745354
Encompass
Mixer HC ISO Rack for 24 x 0.5 mL microcentrifuge tubes, polycarbonate. Includes lid.
Catalog No. 50-274-5354 Supplier USA Scientific Inc Supplier No. 80120020
Mixer HC ISO Rack for 24 x 0.5 mL microcentrifuge tubes, polycarbonate. Includes lid.