Learn More
Arrow Engineering S-316-20 Type 316 Stainless Steel 20" Propeller Shaft
Supplier:  Arrow Engineering S31620
Encompass_Preferred
All Arrow Mixing Products' stainless steel mixer shafts are 3/8" dia. & made of 316 stainless steel. These quality mixer parts have a 5/16-24 external thread for easy assembly with all of our stainless steel propellers and couplings. (Note: propeller and coupling sold separately)
Catalog No. NC2041660