Learn More
Cole-Parmer Cole-Parmer Mixer Testing Beads, Yellow Acetate, 3.0 mm Diameter; 2400 Beads/Jar
Catalog No. NC0646001 Shop All Cole-Parmer Products
Encompass_Preferred
These highly visible beads are calibrated to simulate particles in your fluid and let you easily visualize mixing patterns.
Catalog No. NC0646001 Supplier Cole-Parmer Supplier No. 0456071
These highly visible beads are calibrated to simulate particles in your fluid and let you easily visualize mixing patterns.