Learn More
Perkin Elmer US LLC Mixing/Separation Assembly
Catalog No. 509042497
Encompass_Preferred
Complete modular unit, consisting of: two mixing manifolds with tubing adapters K and M. A gas-liquid separator with a PTFE membrane 5 spare PTFE membranes 1 PTFE tube 110mm long, and 1 PTFE tube 300mm long. Can be used with AA, ICP-OES, ICP-MS and Flow Injection Mercury Systems.
Catalog No. 50-904-2497 Supplier Perkin Elmer US LLC Supplier No. B0507957
Complete modular unit, consisting of: two mixing manifolds with tubing adapters K and M. A gas-liquid separator with a PTFE membrane 5 spare PTFE membranes 1 PTFE tube 110mm long, and 1 PTFE tube 300mm long. Can be used with AA, ICP-OES, ICP-MS and Flow Injection Mercury Systems.