Learn More
Sklar Instruments Mixing/Solution Bowl, Sklar, Stainless Steel. This bowl measures 6-5/8 in. x 2-1/2 in. and has a capacity of 7/8 quart. EA/1.
SDP
Catalog No. 501436995 Shop All Sklar Instruments Products
Encompass_Preferred
Mixing/Solution Bowl, Sklar, Stainless Steel. Sklar Mixing/Solution Bowls are ideal for mixing and containing solutions required in surgical procedures. They may be found in emergency departments where wound cleaning is necessary, or operating rooms where irrigation is required for specific procedures. The top and lip of the mixing bowl does not angle outward such as that of a basin may. This bowl measures 6-5/8 in. x 2-1/2 in. and has a capactiy of 7/8 quart.
This item is not returnable. View return policy
Catalog No. 50-143-6995 Supplier Sklar Instruments Supplier No. 101488
Mixing/Solution Bowl, Sklar, Stainless Steel. Sklar Mixing/Solution Bowls are ideal for mixing and containing solutions required in surgical procedures. They may be found in emergency departments where wound cleaning is necessary, or operating rooms where irrigation is required for specific procedures. The top and lip of the mixing bowl does not angle outward such as that of a basin may. This bowl measures 6-5/8 in. x 2-1/2 in. and has a capactiy of 7/8 quart.