Learn More
Leica Biosystems MIXING STATION
Catalog No. NC0894456
Encompass_Preferred
BOND Mixing Stations are reusable inserts with six vials for mixing and catalyzing chromogens prior to slide application.Fresh chromogen promotes high quality staining.Replacing the mixing stations at recommended intervals ensures that the mixed chromogen does not become contaminated.
Catalog No. NC0894456 Supplier Leica Biosystems Supplier No. S21.1971
BOND Mixing Stations are reusable inserts with six vials for mixing and catalyzing chromogens prior to slide application.Fresh chromogen promotes high quality staining.Replacing the mixing stations at recommended intervals ensures that the mixed chromogen does not become contaminated.