Learn More
Aviva Systems Biology Corp MIXL1 Antibody - N-terminal region (ARP30077_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30077_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MIXL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MIXL1. Alternate names: MIX, MIXL, MILD1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-54115