Learn More
Sklar Instruments Mixter Forceps Right Angle, Sklar, 7-1/4 in., Right Angled (90 Degrees), Longitudinally Serrated, Stainless Steel. EA/1.
SDP
Catalog No. 501437396 Shop All Sklar Instruments Products
Encompass_Preferred
Mixter Forceps Right Angle, Sklar, 7-1/4 in., Right Angled (90 Degrees), Longitudinally Serrated, Stainless Steel. Sklar Mixter Right Angle Forceps are ratcheted, finger ring forceps commonly used for working in obscured surgical sites. They are most frequently used for clamping, dissection, or grasping tissue. Mixter Right Angle Forceps are available in multiple lengths and have serrations the entire length of the jaw. This product has straight shanks, fully serrated jaws, and right angle tips. The length is 7-1/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7396 Supplier Sklar Instruments Supplier No. 171772
Mixter Forceps Right Angle, Sklar, 7-1/4 in., Right Angled (90 Degrees), Longitudinally Serrated, Stainless Steel. Sklar Mixter Right Angle Forceps are ratcheted, finger ring forceps commonly used for working in obscured surgical sites. They are most frequently used for clamping, dissection, or grasping tissue. Mixter Right Angle Forceps are available in multiple lengths and have serrations the entire length of the jaw. This product has straight shanks, fully serrated jaws, and right angle tips. The length is 7-1/4 inches.