Learn More
Chem Service Inc MIXTURE#1 BASE NEUTRALS 1 ML
SDP
Catalog No. NC0806248
Encompass_Preferred
Mixture #1-Base Neutrals, 2000ug/ml in Methylene chloride, Size: 1mL Ampule
Catalog No. NC0806248 Supplier Chem Service Inc Supplier No. MPPHC1X51ML
Mixture #1-Base Neutrals, 2000ug/ml in Methylene chloride, Size: 1mL Ampule