Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mixture Of Metal Scavengers On Silica Gel

Catalog No. 90067310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-067-310G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-067-310G Supplier Sigma Aldrich Supplier No. 90067310G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

It is a proprietary mixture of metal scavengers grafted on silica gel effective in complexing the following: Ca, Cd, Co, Cr, Cs, Cu, Fe, Ir, La, Li, Mg, Ni, Os, Pd, Pt, Rh, Ru, Sc, and Zn.SiliaMetS is a registered trademark of Silicycle, Inc.

Specifications

CAS 74654-06-1
Quantity 100 mg
Synonym Iridium(III) bis[2-(2,4-difluorophenyl)-5-methylpyridine-N,C20]-4,40-di-tert-butyl-2,20-bipyridine hexafluorophosphate
Molecular Weight (g/mol) 1013.97 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.