Learn More
Medchemexpress LLC Mizagliflozin | 666843-10-3 | 99.2% | 564.67 | 5 MG
SDP
Catalog No. 5000387448
Encompass_Preferred
Mizagliflozin (DSP-3235 free base) is a potent, orally active, and selective SGLT1 inhibitor with a Ki of 27 nM for human SGLT1. It shows 303-fold selectivity over SGLT2. Mizagliflozin is used as an antidiabetic agent that can modify postprandial blood glucose excursion. It also exhibits potential in the amelioration of chronic constipation.
  • Potent, orally active, and selective SGLT1 inhibitor (Ki of 27 nM for human SGLT1)
  • 303-fold selectivity over SGLT2
  • Solid appearance, off-white to light yellow color
  • Used as an antidiabetic agent to modify postprandial blood glucose excursion
  • Exhibits potential in the amelioration of chronic constipation
Catalog No. 50-003-87448 Supplier Medchemexpress LLC Supplier No. HY176385MG
Mizagliflozin (DSP-3235 free base) is a potent, orally active, and selective SGLT1 inhibitor with a Ki of 27 nM for human SGLT1. It shows 303-fold selectivity over SGLT2. Mizagliflozin is used as an antidiabetic agent that can modify postprandial blood glucose excursion. It also exhibits potential in the amelioration of chronic constipation.
  • Potent, orally active, and selective SGLT1 inhibitor (Ki of 27 nM for human SGLT1)
  • 303-fold selectivity over SGLT2
  • Solid appearance, off-white to light yellow color
  • Used as an antidiabetic agent to modify postprandial blood glucose excursion
  • Exhibits potential in the amelioration of chronic constipation