Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mizagliflozin (DSP-3235 free base) | 666843-10-3 | 99.2% | 564.67 g/mol | C28H44N4O8 | 10 MG
SDP

Catalog No. 5000298360
Encompass_Preferred

Mizagliflozin is a potent, orally active, selective SGLT1 inhibitor used in research on postprandial glucose regulation and constipation amelioration. It exhibits a Ki of approximately 27 nM for human SGLT1 and about 303-fold selectivity versus SGLT2.

  • Selective SGLT1 inhibition (Ki ≈ 27 nM).
  • High purity (99.2%).
  • Molecular formula C28H44N4O8; molecular weight 564.67 g/mol.
  • Solid, off-white to light yellow appearance.
  • Store at 4°C protected from light; in solution store -80°C for long-term.

Catalog No. 50-002-98360 Supplier Medchemexpress LLC Supplier No. HY1763810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mizagliflozin is a potent, orally active, selective SGLT1 inhibitor used in research on postprandial glucose regulation and constipation amelioration. It exhibits a Ki of approximately 27 nM for human SGLT1 and about 303-fold selectivity versus SGLT2.

  • Selective SGLT1 inhibition (Ki ≈ 27 nM).
  • High purity (99.2%).
  • Molecular formula C28H44N4O8; molecular weight 564.67 g/mol.
  • Solid, off-white to light yellow appearance.
  • Store at 4°C protected from light; in solution store -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.