Learn More
Selleck Chemical LLC Mizagliflozin5mg
SDP
Supplier:  Selleck Chemical LLC S89395MG
Encompass_Preferred
Mizagliflozin (KWA 0711) is a novel, potent, selectivesodium glucose co-transporter 1 (SGLT1) inhibitor with Ki of 27 nM for human SGLT1. The selectivity ratio (Ki value for human SGLT2/Ki value for human SGLT1) of mizagliflozin is 303. Mizagliflozin shows the potential use for the amelioration of chronic constipation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3446