Learn More
Aviva Systems Biology Corp MIZF Antibody - middle region (ARP32543_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32543_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MIZF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MIZF. Alternate names: DKFZP434F162, HiNF-P, ZNF743, MIZFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54646