Learn More
Apexbio Technology LLC Mizolastine 108612-45-9 50mg
SDP
Catalog No. 5000092086
Encompass_Preferred
Mizolastine (CAS 108612-45-9) is a selective antagonist of the histamine H1 receptor exhibiting an inhibitory concentration (IC50) of 47 nM By blocking H1 receptor-mediated signaling Mizolastine effectively attenuates histamine-induced cellular responses involved in allergic inflammation This compound is utilized in biomedical research to investigate mechanisms underlying allergic disorders such as seasonal allergic rhinitis and urticaria and to evaluate therapeutic strategies targeting H1 receptor pathways in allergic and inflammatory conditions
Catalog No. 50-000-92086 Supplier Apexbio Technology LLC Supplier No. B155150
Mizolastine (CAS 108612-45-9) is a selective antagonist of the histamine H1 receptor exhibiting an inhibitory concentration (IC50) of 47 nM By blocking H1 receptor-mediated signaling Mizolastine effectively attenuates histamine-induced cellular responses involved in allergic inflammation This compound is utilized in biomedical research to investigate mechanisms underlying allergic disorders such as seasonal allergic rhinitis and urticaria and to evaluate therapeutic strategies targeting H1 receptor pathways in allergic and inflammatory conditions