Learn More
Medchemexpress LLC Mizolastine | 108612-45-9 | 99.8% | 25 MG
SDP
Catalog No. 5000406524
Encompass_Preferred
Mizolastine is an orally active, high affinity, and specific peripheral histamine H1 receptor antagonist. It effectively inhibits mRNA expression of VEGF165, VEGF120, TNF-α, and KC, and can be used in studies of allergic rhinitis and chronic idiopathic urticaria.
  • Specific peripheral histamine H1 receptor antagonist
  • Inhibits mRNA expression of VEGF165, VEGF120, TNF-α, and KC
  • Suitable for research on allergic rhinitis
  • Suitable for research on chronic idiopathic urticaria
  • High purity of 99.8%
Catalog No. 50-004-06524 Supplier Medchemexpress LLC Supplier No. 25MGHYB0164
Mizolastine is an orally active, high affinity, and specific peripheral histamine H1 receptor antagonist. It effectively inhibits mRNA expression of VEGF165, VEGF120, TNF-α, and KC, and can be used in studies of allergic rhinitis and chronic idiopathic urticaria.
  • Specific peripheral histamine H1 receptor antagonist
  • Inhibits mRNA expression of VEGF165, VEGF120, TNF-α, and KC
  • Suitable for research on allergic rhinitis
  • Suitable for research on chronic idiopathic urticaria
  • High purity of 99.8%