Learn More
TARGETMOL CHEMICALS INC Mizolastine 200MG
SDP
Catalog No. 502880543
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Mizolastine (SL 850324) (IC50= 47 nM) is a histamine H1-receptor antagonist.It is used in the treatment of allergic reactions such as hay fever (seasonal allergic rhinitis), hives. Purity 99.77%
Catalog No. 50-288-0543 Supplier TARGETMOL CHEMICALS INC Supplier No. T0118200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Mizolastine (SL 850324) (IC50= 47 nM) is a histamine H1-receptor antagonist.It is used in the treatment of allergic reactions such as hay fever (seasonal allergic rhinitis), hives. Purity 99.77%