Learn More
Selleck Chemical LLC Mizolastine S2107-50mg
SDP
Catalog No. 503109696
Encompass_Preferred
Mizolastine (SL 850324) is a histamine H1-receptor antagonist with IC50 of 47 nM used in the treatment of hay fever (seasonal allergic rhinitis) hives and other allergic reactions
Catalog No. 50-310-9696 Supplier Selleck Chemical LLC Supplier No. S210750MG
Mizolastine (SL 850324) is a histamine H1-receptor antagonist with IC50 of 47 nM used in the treatment of hay fever (seasonal allergic rhinitis) hives and other allergic reactions