Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mizolastine 98.0+%, TCI America™
SDP

Catalog No. M27561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27561G Supplier TCI America Supplier No. M27561G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108612-45-9
Molecular Formula C24H25FN6O
Molecular Weight (g/mol) 432.50
MDL Number MFCD00864625
InChI Key PVLJETXTTWAYEW-UHFFFAOYSA-N
PubChem CID 65906
IUPAC Name 2-[(1-{1-[(4-fluorophenyl)methyl]-1H-1,3-benzodiazol-2-yl}piperidin-4-yl)(methyl)amino]-3,4-dihydropyrimidin-4-one
SMILES CN(C1CCN(CC1)C1=NC2=CC=CC=C2N1CC1=CC=C(F)C=C1)C1=NC=CC(=O)N1

Specifications

Melting Point 224°C
Color White
Quantity 1 g
Formula Weight 432.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mizolastine
RTECSNumber UW7481284
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.