Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MJ33 lithium salt 10mg | 1007476-63-2 | 10 MG
SDP

Catalog No. 5000228478
Encompass_Preferred

MJ33 lithium salt is the lithium salt form of MJ33, an active-site-directed, specific, competitive, and reversible phospholipase A2 (PLA2) inhibitor that blocks the calcium-independent PLA2 (iPLA2) activity of Peroxiredoxin-6 (Prdx6). It is supplied as a high-purity research reagent for biochemical and cell-signaling studies.

  • Specific, competitive, reversible inhibitor of PLA2 that blocks iPLA2 activity of Prdx6.
  • High purity suitable for research applications (reported purity 99.0%).
  • Available in small research quantities for in vitro and in vivo assays.
  • Molecular weight 498.48 and formula C22H43F3LiO6P for precise dosing and formulation.
  • Recommended storage: -20°C, sealed and away from moisture; in solvent storage conditions provided.

Catalog No. 50-002-28478 Supplier Medchemexpress LLC Supplier No. HY11506210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MJ33 lithium salt is the lithium salt form of MJ33, an active-site-directed, specific, competitive, and reversible phospholipase A2 (PLA2) inhibitor that blocks the calcium-independent PLA2 (iPLA2) activity of Peroxiredoxin-6 (Prdx6). It is supplied as a high-purity research reagent for biochemical and cell-signaling studies.

  • Specific, competitive, reversible inhibitor of PLA2 that blocks iPLA2 activity of Prdx6.
  • High purity suitable for research applications (reported purity 99.0%).
  • Available in small research quantities for in vitro and in vivo assays.
  • Molecular weight 498.48 and formula C22H43F3LiO6P for precise dosing and formulation.
  • Recommended storage: -20°C, sealed and away from moisture; in solvent storage conditions provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.