Learn More
Medchemexpress LLC HY-117474 10mg Medchemexpress, MJN110 CAS:1438416-21-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11747410MG
Encompass_Preferred
Medchemexpress, HY-117474 10mg MJN110 CAS:1438416-21-7 MJN110 is an orally active and selective monoacylglycerol lipase (MAGL) inhibitor with IC50s of 9.1 nM and 2.1 nM for hMAGL and 2-arachidonoylglycerol (2-AG), respectively. MJN110 produces opioid-sparing effects and displays strong antihyperalgesic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9475