Learn More
ATCC MJ; T Cell Lymphoma; Human (Homo sapiens)”
Catalog No. 502384828
Encompass
MJ [G11] is a T lymphoblast cell that was isolated from the peripheral blood of a 50-year-old, White male with cutaneous T cell lymphoma. The cell can be used in immuno-oncology research.
Catalog No. 50-238-4828 Supplier ATCC Supplier No. CRL8294
MJ [G11] is a T lymphoblast cell that was isolated from the peripheral blood of a 50-year-old, White male with cutaneous T cell lymphoma. The cell can be used in immuno-oncology research.