Learn More
Medchemexpress LLC HY-U00076 1mg Medchemexpress, MK-0249 CAS:862309-06-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU000761MG
Encompass_Preferred
Medchemexpress, HY-U00076 1mg MK-0249 CAS:862309-06-6 MK-0249 is a potent histamine H3 receptor antagonist, with Ki of 1.7 nM for human H3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0620